Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Cynomolgus Rhesus macaque CTLA-4, Avitag,His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus maca 1 * 25 µG
Catalog # ACRBCT4-C82E5-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus Rhesus macaque CTLA-4, Avitag,His Tag (CT4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Ser 160 (Accession G7PL88-1). In the region Ala 37 - Ser 160, the AA sequence of Cynomolgus and Rhesus maca 1 * 25 µG
Catalog # ACRBCT4-C82E5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together