Order Entry
Netherlands
ContactUsLinkComponent
MABSol® Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession NP001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). null
Catalog # ACRBER3-H8223200UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Human ErbB3, His Tag, primary amine labeling (ER3-H8223) is expressed from human HEK293 cells. It contains AA Ser 20 - Thr 643 (Accession NP001973.2). It is the biotinylated form of Human ErbB3, His Tag (ER3-H5223). null
Catalog # ACRBER3-H8223200UG
Supplier:  ACRO Biosystems
Frequently Bought Together