Order Entry
Netherlands
ContactUsLinkComponent
MABSol® Biotinylated Human GM-CSF (GMF-H8214) is expressed from human HEK293 cells. It contains AA Ala 18 - Glu 144 (Accession NP00 0749.2) It is the biotinylated form of ActiveMax® Human GM-CSF (Cat GMF-H4214). null
Catalog # ACRBGMF-H8214-25UG
Supplier:  ACRO Biosystems
MABSol® Biotinylated Human GM-CSF (GMF-H8214) is expressed from human HEK293 cells. It contains AA Ala 18 - Glu 144 (Accession NP00 0749.2) It is the biotinylated form of ActiveMax® Human GM-CSF (Cat GMF-H4214). null
Catalog # ACRBGMF-H8214-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together