Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Biotinylated Human LILRA3 CD85e Protein, His,Avitag™,Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is express ed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession AAH28208.1).This protein carries a polyh 1 * 200 µG
Catalog # ACRBLI3-H82E0200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human LILRA3 CD85e Protein, His,Avitag™,Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is express ed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession AAH28208.1).This protein carries a polyh 1 * 200 µG
Catalog # ACRBLI3-H82E0200UG
Supplier:  ACRO Biosystems
Frequently Bought Together