Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Human SLAMF7, Avitag,His Tag (SL7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 ( Accession Q9NQ25-1 null
Catalog # ACRBSL7-H82E0-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Human SLAMF7, Avitag,His Tag (SL7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ser 23 - Met 226 ( Accession Q9NQ25-1 null
Catalog # ACRBSL7-H82E0-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together