Order Entry
Netherlands
ContactUsLinkComponent
MABSol® Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gl n 24 - Tyr 213 (Accession AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245). 1 * 200 µG
  ACRBSOT-H8245200UG
MABSol® Biotinylated Human SOST, His Tag, primary amine labeling (SOT-H8245) is expressed from human HEK293 cells. It contains AA Gl n 24 - Tyr 213 (Accession AAI01087.1). It is the biotinylated form of Human SOST, His Tag (HST-H5245). 1 * 200 µG
  ACRBSOT-H8245200UG
Frequently Bought Together