Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Avitag™,His Tag,Biotinylated Human EpCAM, Avitag,His Tag (EPM-H82E8) is e xpressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a 1 * 200 µG
Catalog # ACRBEPM-H82E8200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human EpCAM TROP1 Protein, Avitag™,His Tag,Biotinylated Human EpCAM, Avitag,His Tag (EPM-H82E8) is e xpressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession AAH14785.1).This protein carries a 1 * 200 µG
Catalog # ACRBEPM-H82E8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together