Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Biotinylated Human B7-H5 Gi24 VISTA Protein, Fc,Avitag™,Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expres sed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568.1).This protein carries a huma 1 * 25 µG
Catalog # ACRBB75-H82F3-25UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human B7-H5 Gi24 VISTA Protein, Fc,Avitag™,Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expres sed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession AAH20568.1).This protein carries a huma 1 * 25 µG
Catalog # ACRBB75-H82F3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together