Order Entry
Netherlands
ContactUsLinkComponent
Cynomolgus Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Acce ssion G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus. 1 * 100 µG
Catalog # ACRBCD6-C5254100UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque B7-2, Fc Tag (CD6-C5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Acce ssion G7NXR4). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7-2 are homologus. 1 * 100 µG
Catalog # ACRBCD6-C5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together