Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Cynomolgus CD3 delta Protein, His Tag,Cynomolgus CD3 delta, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession Q95LI8).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG
Catalog # ACRBCDD-C52H8100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Cynomolgus CD3 delta Protein, His Tag,Cynomolgus CD3 delta, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession Q95LI8).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG
Catalog # ACRBCDD-C52H8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together