Bestelling invoeren
Netherlands
ContactUsLinkComponent
Cynomolgus Rhesus macaque IL-4 R alpha, His Tag (ILR-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 26 - Arg 232 (Accession G7Q0S7). In the region Met 26 - Arg 232, the AA sequence of Cynomolgus and Rhesus macaque IL-4 R alph s 1 * 1 EA
Catalog # ACRBILR-C52H8-50UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque IL-4 R alpha, His Tag (ILR-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 26 - Arg 232 (Accession G7Q0S7). In the region Met 26 - Arg 232, the AA sequence of Cynomolgus and Rhesus macaque IL-4 R alph s 1 * 1 EA
Catalog # ACRBILR-C52H8-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together