Order Entry
Netherlands
ContactUsLinkComponent
Cynomolgus Rhesus macaque PD-L1, His Tag (PD1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Ac cession G7PSE7-1). In the region Phe 19 - Arg 238, the AA sequence of Cynomolgus and Rhesus macaque PD-L1 are homolog 1 * 100 µG
Catalog # ACRBPD1-C52H4100UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque PD-L1, His Tag (PD1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Ac cession G7PSE7-1). In the region Phe 19 - Arg 238, the AA sequence of Cynomolgus and Rhesus macaque PD-L1 are homolog 1 * 100 µG
Catalog # ACRBPD1-C52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together