Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse LILRB4 CD85k ILT3 Protein, His Tag,Mouse LILRB4, His Tag (CDK-M5229) is expressed from human 293 cells (HEK 293). It contains AA Gly 24 - Lys 238 (Accession Q64281-1 ).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg
Catalog # ACRBCDK-M5229-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse LILRB4 CD85k ILT3 Protein, His Tag,Mouse LILRB4, His Tag (CDK-M5229) is expressed from human 293 cells (HEK 293). It contains AA Gly 24 - Lys 238 (Accession Q64281-1 ).This protein carries a polyhistidine tag at the C-terminu 1 * 1 mg
Catalog # ACRBCDK-M5229-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together