Order Entry
Netherlands
ContactUsLinkComponent
Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (B71-H52A4) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 24 2 (Accession P33681-1). 1 * 100 µG
Catalog # ACRBB71-H52A4100UG
Supplier:  ACRO Biosystems
Human B7-1, Mouse IgG2a Fc Tag, low endotoxin (B71-H52A4) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 24 2 (Accession P33681-1). 1 * 100 µG
Catalog # ACRBB71-H52A4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together