Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Axl Protein, His Tag,Human Axl, His Tag (AXL-H5226) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 449 (Accession AAH32229).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBAXL-H5226-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Axl Protein, His Tag,Human Axl, His Tag (AXL-H5226) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 449 (Accession AAH32229).This protein carries a polyhistidine tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBAXL-H5226-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together