Recombinat Protein,Rhesus macaque B7-H5 Gi24 VISTA Protein, His Tag,Rhesus macaque B7-H5, His Tag (B75-C5220) is expressed from huma n 293 cells (HEK293). It contains AA Phe 33 - Thr 192 (Accession NP001177807.1).This protein carries a polyhistidine 1 * 100 µG
Catalog # ACRBB75-C5220100UG
Supplier: ACRO Biosystems
Frequently Bought Together