Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human VSIG8 Protein, Fc tag,Human VSIG8, Fc Tag (VS8-H5250) is expressed from human 293 cells (HEK293). It contai ns AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 100 µG
Catalog # ACRBVS8-H5250100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human VSIG8 Protein, Fc tag,Human VSIG8, Fc Tag (VS8-H5250) is expressed from human 293 cells (HEK293). It contai ns AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 100 µG
Catalog # ACRBVS8-H5250100UG
Supplier:  ACRO Biosystems
Frequently Bought Together