Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human BACE-1 Protein, Fc Tag,Human BACE-1, Fc Tag (BA1-H5261) is expressed from human 293 cells (HEK293). It cont ains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries a human IgG1 Fc tag at the C-terminus. The prote 1 * 200 µG
Catalog # ACRBBA1-H5261200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human BACE-1 Protein, Fc Tag,Human BACE-1, Fc Tag (BA1-H5261) is expressed from human 293 cells (HEK293). It cont ains AA Thr 22 - Thr 457 (Accession NP036236.1).This protein carries a human IgG1 Fc tag at the C-terminus. The prote 1 * 200 µG
Catalog # ACRBBA1-H5261200UG
Supplier:  ACRO Biosystems
Frequently Bought Together