Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBAL3-H5259-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human BMPR-IA ALK-3 Protein, Fc Tag,Human BMPR-IA, Fc Tag (AL3-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Arg 152 (Accession CAA80257).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 1 mg
Catalog # ACRBAL3-H5259-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together