Recombinat Protein,Human VSIG8 Protein, His Tag,Human VSIG8, His Tag (VS8-H52H1) is expressed from human 293 cells (HEK293). It cont ains AA Val 22 - Gly 263 (Accession NP001013683.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 100 µG
Catalog # ACRBVS8-H52H1100UG
Supplier: ACRO Biosystems
Frequently Bought Together