Order Entry
Netherlands
ContactUsLinkComponent
Cynomolgus TIM-3, Fc Tag (TI3-C525b) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 201 (Accession EHH54703 .1). 1 * 1 mg
Catalog # ACRBTI3-C525B-1MG
Supplier:  ACRO Biosystems
Cynomolgus TIM-3, Fc Tag (TI3-C525b) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 201 (Accession EHH54703 .1). 1 * 1 mg
Catalog # ACRBTI3-C525B-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together