Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD14 Protein, His Tag,Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Met 344 (Accession AAH10507).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD4-H5228100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD14 Protein, His Tag,Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Met 344 (Accession AAH10507).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD4-H5228100UG
Supplier:  ACRO Biosystems
Frequently Bought Together