Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human CD163 Protein, His Tag,Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It cont ains AA Ser 42 - Ser 1045 (Accession Q86VB7).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBCD3-H52E8100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD163 Protein, His Tag,Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It cont ains AA Ser 42 - Ser 1045 (Accession Q86VB7).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBCD3-H52E8100UG
Supplier:  ACRO Biosystems
Frequently Bought Together