Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Biotinylated Human BTN3A1 CD277 Protein, Fc,Avitag™,Biotinylated Human BTN3A1, Fc,Avitag (BT1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession O00481-1).This protein carries a human IgG 1 * 200 µG
Catalog # ACRBBT1-H82F7200UG
Supplier:  ACRO Biosystems
Recombinat Protein,Biotinylated Human BTN3A1 CD277 Protein, Fc,Avitag™,Biotinylated Human BTN3A1, Fc,Avitag (BT1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Gly 254 (Accession O00481-1).This protein carries a human IgG 1 * 200 µG
Catalog # ACRBBT1-H82F7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together