Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Unconjugated Human TPBG 5T4 Protein, His,Avitag,Unconjugated Human TPBG, His,Avitag (TPG-H52E5) is expressed fro m human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidine 1 * 1 mg
Catalog # ACRBTPG-H52E5-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Unconjugated Human TPBG 5T4 Protein, His,Avitag,Unconjugated Human TPBG, His,Avitag (TPG-H52E5) is expressed fro m human 293 cells (HEK293). It contains AA Ser 32 - Ser 355 (Accession Q13641-1).This protein carries a polyhistidine 1 * 1 mg
Catalog # ACRBTPG-H52E5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together