Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Rhesus macaque EpCAM TROP1 Protein, His Tag,Rhesus macaque EpCAM, His Tag (EPM-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession Q1WER1).This protein carries a polyhistidine tag at the
Catalog # ACRBEPM-C5227-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Rhesus macaque EpCAM TROP1 Protein, His Tag,Rhesus macaque EpCAM, His Tag (EPM-C5227) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession Q1WER1).This protein carries a polyhistidine tag at the
Catalog # ACRBEPM-C5227-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together