Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human TrkB NTRK2 Protein, Fc Tag,Human TrkB, Fc Tag (NT2-H5254) is expressed from human 293 cells (HEK293). It co ntains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a human IgG1 Fc tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBNT2-H5254100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human TrkB NTRK2 Protein, Fc Tag,Human TrkB, Fc Tag (NT2-H5254) is expressed from human 293 cells (HEK293). It co ntains AA Cys 32 - His 430 (Accession AAH31835).This protein carries a human IgG1 Fc tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBNT2-H5254100UG
Supplier:  ACRO Biosystems
Frequently Bought Together