Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Llama IgG2b Fc Tag, low endotoxin,Human Mucin-1, Llama IgG2b Fc Tag, low endotoxin ( MU1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession AAI20976).This prot 1 * 100 µG
Catalog # ACRBMU1-H5255100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Llama IgG2b Fc Tag, low endotoxin,Human Mucin-1, Llama IgG2b Fc Tag, low endotoxin ( MU1-H5255) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession AAI20976).This prot 1 * 100 µG
Catalog # ACRBMU1-H5255100UG
Supplier:  ACRO Biosystems
Frequently Bought Together