Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human GFR alpha-like Protein, His Tag,Human GFR alpha-like, His Tag (GFA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Glu 351 (Accession Q6UXV0-1).This protein carries a polyhistidine tag at the C-ter 1 * 100 µG
Catalog # ACRBGFA-H52H3100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human GFR alpha-like Protein, His Tag,Human GFR alpha-like, His Tag (GFA-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Glu 351 (Accession Q6UXV0-1).This protein carries a polyhistidine tag at the C-ter 1 * 100 µG
Catalog # ACRBGFA-H52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together