Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The p 1 * 1 mg
Catalog # ACRBNKA-H5244-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human NKG2A CD159a Protein, His Tag,Human NKG2A, His Tag (NKA-H5244) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession P26715-1).This protein carries a polyhistidine tag at the N-terminus. The p 1 * 1 mg
Catalog # ACRBNKA-H5244-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together