Recombinat Protein,Human Siglec-8 Protein, Fc Tag,Human Siglec-8, Fc Tag (SI8-H5254) is expressed from human 293 cells (HEK293). It contains AA Met 17 - Ala 363 (Accession Q9NYZ4-1).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 100 µG
Catalog # ACRBSI8-H5254100UG
Supplier: ACRO Biosystems
Frequently Bought Together