Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBMU1-H5252100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Mucin-1 MUC-1 Protein, Fc Tag,Human Mucin-1, Fc Tag (MU1-H5252) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Gly 167 (Accession AAI20976).This protein carries a human IgG1 Fc tag at the C-terminus. The 1 * 100 µG
Catalog # ACRBMU1-H5252100UG
Supplier:  ACRO Biosystems
Frequently Bought Together