Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human LRRC4 Protein, His Tag,Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 39 - Lys 527 (Accession AAI11562.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBLR4-H5221100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human LRRC4 Protein, His Tag,Human LRRC4, His Tag (LR4-H5221) is expressed from human 293 cells (HEK293). It cont ains AA Ala 39 - Lys 527 (Accession AAI11562.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBLR4-H5221100UG
Supplier:  ACRO Biosystems
Frequently Bought Together