Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,HCMV Glycoprotein B (gB) Protein, Fc Tag,HCMV Glycoprotein B (gB), Fc Tag (CMB-V5255) Val 23 - Lys 700 (Accession P13201-1) with furin cleavage site mutated from RTKR to TTQT, was produced in human 293 cells (HEK293) at ACROBiosys 1 * 50 µG
Catalog # ACRBCMB-V5255-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,HCMV Glycoprotein B (gB) Protein, Fc Tag,HCMV Glycoprotein B (gB), Fc Tag (CMB-V5255) Val 23 - Lys 700 (Accession P13201-1) with furin cleavage site mutated from RTKR to TTQT, was produced in human 293 cells (HEK293) at ACROBiosys 1 * 50 µG
Catalog # ACRBCMB-V5255-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together