Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 50 µG
Catalog # ACRBDK3-M5226-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse Dkk-3 Protein, His Tag,Mouse Dkk-3, His Tag (DK3-M5226) is expressed from human 293 cells (HEK293). It cont ains AA Pro 23 - Ile 349 (Accession NP056629.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 50 µG
Catalog # ACRBDK3-M5226-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together