Recombinat Protein,Human Siglec-5 CD170 Protein, Fc Tag,Human Siglec-5, Fc Tag (SI5-H5250) is expressed from human 293 cells (HEK293 ). It contains AA Glu 17 - Thr 434 (Accession AAH29896).This protein carries a human IgG1 Fc tag at the C-terminus. T 1 * 100 µG
Catalog # ACRBSI5-H5250100UG
Supplier: ACRO Biosystems
Frequently Bought Together