Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells (H EK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Catalog # ACRBFZ7-H52H9100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Frizzled-7 FZD7 Protein, His Tag,Human Frizzled-7, His Tag (FZ7-H52H9) is expressed from human 293 cells (H EK293). It contains AA Gln 33 - Leu 185 (Accession O75084-1).This protein carries a polyhistidine tag at the C-termin 1 * 100 µG
Catalog # ACRBFZ7-H52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together