Order Entry
Netherlands
ContactUsLinkComponent
Human PD-1, Fc Tag, low endotoxin (HPLC-verified) (PD1-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gl n 167 (Accession NP005009.2). 1 * 1 mg
Catalog # ACRBPD1-H5257-1MG
Supplier:  ACRO Biosystems
Human PD-1, Fc Tag, low endotoxin (HPLC-verified) (PD1-H5257) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gl n 167 (Accession NP005009.2). 1 * 1 mg
Catalog # ACRBPD1-H5257-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together