Order Entry
Netherlands
ContactUsLinkComponent
Human ITGA10ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Ser 1122 (ITGA10) Gln 21 - Asp 728 (ITGB1) (Accession O75578-1 (ITGA10) P05556-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-H52WA100UG
Supplier:  ACRO Biosystems
Human ITGA10ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52Wa) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Ser 1122 (ITGA10) Gln 21 - Asp 728 (ITGB1) (Accession O75578-1 (ITGA10) P05556-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-H52WA100UG
Supplier:  ACRO Biosystems
Frequently Bought Together