Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,FITC-Labeled Human PSMA FOLH1 Protein, His Tag,FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from hum an 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1). It is the FITC labeled form of Human PSM 1 * 200 µG
Catalog # ACRBPSA-HF244200UG
Supplier:  ACRO Biosystems
Recombinat Protein,FITC-Labeled Human PSMA FOLH1 Protein, His Tag,FITC-Labeled Human PSMA, His Tag (PSA-HF244) is expressed from hum an 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession Q04609-1). It is the FITC labeled form of Human PSM 1 * 200 µG
Catalog # ACRBPSA-HF244200UG
Supplier:  ACRO Biosystems
Frequently Bought Together