Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Rhesus macaque SLAMF7 CRACC CD319 Protein, His Tag,Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from h uman 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession F7HQ72-1).This protein carries a polyhistidine ta 1 * 1 mg
Catalog # ACRBSL7-R52H7-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Rhesus macaque SLAMF7 CRACC CD319 Protein, His Tag,Rhesus macaque SLAMF7, His Tag (SL7-R52H7) is expressed from h uman 293 cells (HEK293). It contains AA Ser 23 - Met 226 (Accession F7HQ72-1).This protein carries a polyhistidine ta 1 * 1 mg
Catalog # ACRBSL7-R52H7-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together