Order Entry
Netherlands
ContactUsLinkComponent
Human ITGA2ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W6) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Thr 1132 (ITGA2) Gln 21 - Asp 728 (ITGB1) (Accession P17301-1 (ITGA2) P05556-1 (ITGB1)). 1 * 1 mg
Catalog # ACRBIT1-H52W6-1MG
Supplier:  ACRO Biosystems
Human ITGA2ITGB1 Heterodimer Protein, His TagTag Free (IT1-H52W6) is expressed from human 293 cells (HEK293). It contains AA Tyr 30 - Thr 1132 (ITGA2) Gln 21 - Asp 728 (ITGB1) (Accession P17301-1 (ITGA2) P05556-1 (ITGB1)). 1 * 1 mg
Catalog # ACRBIT1-H52W6-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together