Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Fetuin A FETUA Protein, His Tag,Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK2 93). It contains AA Ala 19 - Val 367 (Accession NP001613).This protein carries a polyhistidine tag at the C-terminus. 1 * 1 mg
Catalog # ACRBFEA-H5221-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Fetuin A FETUA Protein, His Tag,Human Fetuin A, His Tag (FEA-H5221) is expressed from human 293 cells (HEK2 93). It contains AA Ala 19 - Val 367 (Accession NP001613).This protein carries a polyhistidine tag at the C-terminus. 1 * 1 mg
Catalog # ACRBFEA-H5221-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together