Order Entry
Netherlands
ContactUsLinkComponent
Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It conta ins AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region Ala 1 * 50 µG
Catalog # ACRBFCM-C52W9-50UG
Supplier:  ACRO Biosystems
Cynomolgus Rhesus macaque FCGRTB2M Heterodimer Protein, His Tag (BLI verified), is expressed from human 293 cells (HEK293). It conta ins AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In the region Ala 1 * 50 µG
Catalog # ACRBFCM-C52W9-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together