Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified), is expressed from human 293 cells (HEK29 3). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In th 1 * 25 µG
Catalog # ACRBFCM-C82W3-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus Rhesus macaque FcRn Heterodimer Protein, His,Avitag (SPR verified), is expressed from human 293 cells (HEK29 3). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession Q8SPV9-1 (FCGRT) Q8SPW0-1 (B2M)). In th 1 * 25 µG
Catalog # ACRBFCM-C82W3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together