Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human GPA33 A33 Protein, His Tag,Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession AAH74830).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 50 µG
Catalog # ACRBGP3-H5224-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human GPA33 A33 Protein, His Tag,Human GPA33, His Tag (GP3-H5224) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession AAH74830).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 50 µG
Catalog # ACRBGP3-H5224-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together