Order Entry
Netherlands
ContactUsLinkComponent
Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 100 µG
Catalog # ACRBTFR-C524A100UG
Supplier:  ACRO Biosystems
Cynomolgus Transferrin R, His Tag (TFR-C524a) is expressed from human 293 cells (HEK293). It contains AA Cys 89 - Phe 760 (Accession XP005545315.1). 1 * 100 µG
Catalog # ACRBTFR-C524A100UG
Supplier:  ACRO Biosystems
Frequently Bought Together