Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK 293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG
Catalog # ACRBFZ5-H5259100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human Frizzled-5 FZD5 Protein, Fc Tag,Human Frizzled-5, Fc Tag (FZ5-H5259) is expressed from human 293 cells (HEK 293). It contains AA Ala 27 - Pro 167 (Accession Q13467).This protein carries a human IgG1 Fc tag at the C-terminus. 1 * 100 µG
Catalog # ACRBFZ5-H5259100UG
Supplier:  ACRO Biosystems
Frequently Bought Together