Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a calc 1 * 1 mg
Catalog # ACRBS10-H5127-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human S100A10 Protein, His Tag,Human S100A10, His Tag (S10-H5127) is expressed from E.coli cells. It contains AA Met 1 - Lys 97 (Accession AAH15973).This protein carries a polyhistidine tag at the C-terminus. The protein has a calc 1 * 1 mg
Catalog # ACRBS10-H5127-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together