Order Entry
Netherlands
ContactUsLinkComponent
Recombinat Protein,Mouse CD73 NT5E Protein, His Tag,Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It c ontains AA Trp 29 - Ser 551 (Accession Q61503-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 500 µG
Catalog # ACRBCD3-M52H9500UG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse CD73 NT5E Protein, His Tag,Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It c ontains AA Trp 29 - Ser 551 (Accession Q61503-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 500 µG
Catalog # ACRBCD3-M52H9500UG
Supplier:  ACRO Biosystems
Frequently Bought Together